COMH93D5D1DD Combi-Blocks, Inc.

2-Chloro-n-(3,5-difluorophenyl)propanamide

  • MDL number MFCD09736935
  • Molecular Weight : 219.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-0427

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-0427
Availability details for
Quantity
  • Availability
  • Add to Cart