COMH93D5D1EE Combi-Blocks, Inc.

5,7-Difluoroindoline-2,3-dione

  • MDL number MFCD09741498
  • Cas Number : 116570-41-3
  • Molecular Weight : 183.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-0445

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-0445
Availability details for
Quantity
  • Availability
  • Add to Cart