COMH93D5D253 Combi-Blocks, Inc.

1-(3-Amino-4-(benzyloxy)phenyl)ethanone

  • MDL number MFCD09753795
  • Cas Number : 14347-15-0
  • Molecular Weight : 241.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-0572

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-0572
Availability details for
Quantity
  • Availability
  • Add to Cart