COMH93D5D2D3 Combi-Blocks, Inc.

3-Methyl-3-azabicyclo[3.3.1]nonan-9-ol

  • MDL number MFCD09834348
  • Cas Number : 13962-79-3
  • Molecular Weight : 155.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-0722

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-0722
Availability details for
Quantity
  • Availability
  • Add to Cart