COMH93D5D2D7 Combi-Blocks, Inc.

2-Amino-1-(5-amino-1h-imidazol-4-yl)ethanone dihydrochloride

  • MDL number MFCD09834711
  • Cas Number : 69195-92-2
  • Molecular Weight : 213.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-0727

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-0727
Availability details for
Quantity
  • Availability
  • Add to Cart