COMH93D5D2ED Combi-Blocks, Inc.

Methyl 6-cyano-1h-indole-3-carboxylate

  • MDL number MFCD09836007
  • Cas Number : 1357147-38-6
  • Molecular Weight : 200.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-0754

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-0754
Availability details for
Quantity
  • Availability
  • Add to Cart