COMH93D5D2F0 Combi-Blocks, Inc.

Imipramine n-oxide monohydrate

  • MDL number MFCD09836207
  • Molecular Weight : 314.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-0757

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-0757
Availability details for
Quantity
  • Availability
  • Add to Cart