COMH93D5D31D Combi-Blocks, Inc.

4-Desmethoxypropoxyl-4-chloro rabeprazole

  • MDL number MFCD09840357
  • Cas Number : 168167-42-8
  • Molecular Weight : 305.79

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-0806

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-0806
Availability details for
Quantity
  • Availability
  • Add to Cart