COMH93D5D32D Combi-Blocks, Inc.

(S)-2-((tert-Butoxycarbonyl)amino)-2,3-dimethylbutanoic acid

  • MDL number MFCD09841839
  • Cas Number : 53940-90-2
  • Molecular Weight : 231.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-0828

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-0828
Availability details for
Quantity
  • Availability
  • Add to Cart