COMH93D5D32E Combi-Blocks, Inc.

Rhodamine 700

  • MDL number MFCD09841902
  • Cas Number : 63561-42-2
  • Molecular Weight : 538.95

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-0829

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-0829
Availability details for
Quantity
  • Availability
  • Add to Cart