COMH93D5D381 Combi-Blocks, Inc.

2-Bromo-1-chloro-4-(difluoromethoxy)benzene

  • MDL number MFCD09878208
  • Cas Number : 1000574-90-2
  • Molecular Weight : 257.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-0932

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-0932
Availability details for
Quantity
  • Availability
  • Add to Cart