COMH93D5D38C Combi-Blocks, Inc.

tert-Butyl 3-(dimethylamino)piperidine-1-carboxylate

  • MDL number MFCD09878592
  • Cas Number : 1000576-83-9
  • Molecular Weight : 228.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-0945

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-0945
Availability details for
Quantity
  • Availability
  • Add to Cart