COMH93D5D38F Combi-Blocks, Inc.

1-Pyrrolidin-2-(3'-chlorophenyl)-2-(n-cbz-n-methyl)amino-ethane hcl

  • MDL number MFCD09878853
  • Molecular Weight : 409.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-0949

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-0949
Availability details for
Quantity
  • Availability
  • Add to Cart