COMH93D5D3B6 Combi-Blocks, Inc.

5-Amino-n,n-dibutyl-2-methyl-benzenesulfonamide

  • MDL number MFCD09901906
  • Molecular Weight : 298.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-0994

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-0994
Availability details for
Quantity
  • Availability
  • Add to Cart