COMH93D5D59F Combi-Blocks, Inc.

N-(2-Bromoethyl)-n-methylaniline

  • MDL number MFCD10697851
  • Cas Number : 51905-47-6
  • Molecular Weight : 214.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-1588

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-1588
Availability details for
Quantity
  • Availability
  • Add to Cart