COMH93D5D5C3 Combi-Blocks, Inc.

5-Bromo-2-fluoro-n-(4-(trifluoromethoxy)phenyl)benzamide

  • MDL number MFCD10915986
  • Cas Number : 1042839-65-5
  • Molecular Weight : 378.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-1631

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-1631
Availability details for
Quantity
  • Availability
  • Add to Cart