COMH93D5D5FA Combi-Blocks, Inc.

4-(Isoquinolin-3-yl)phenol

  • MDL number MFCD11042297
  • Cas Number : 884500-88-3
  • Molecular Weight : 221.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-1698

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-1698
Availability details for
Quantity
  • Availability
  • Add to Cart