COMH93D5D60D Combi-Blocks, Inc.

Ethyl 2,6-difluoro-3-nitrobenzoate

  • MDL number MFCD11042827
  • Cas Number : 150368-36-8
  • Molecular Weight : 231.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-1722

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-1722
Availability details for
Quantity
  • Availability
  • Add to Cart