COMH93D5D63D Combi-Blocks, Inc.

2-Chloro-3-methylbenzylamine

  • MDL number MFCD11045179
  • Cas Number : 1044256-78-1
  • Molecular Weight : 155.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-1784

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-1784
Availability details for
Quantity
  • Availability
  • Add to Cart