COMH93D5D645 Combi-Blocks, Inc.

2-Bromo-5-nitrobenzylamine hydrochloride

  • MDL number MFCD11045215
  • Cas Number : 2301069-19-0
  • Molecular Weight : 267.51

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-1793

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-1793
Availability details for
Quantity
  • Availability
  • Add to Cart