COMH93D5D653 Combi-Blocks, Inc.

Diethyl-12-bromododecylphosphonate

  • MDL number MFCD11046105
  • Cas Number : 264231-28-9
  • Molecular Weight : 385.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-1811

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-1811
Availability details for
Quantity
  • Availability
  • Add to Cart