COMH93D5D6FF Combi-Blocks, Inc.

Deoxynojirimycin tetrabenzyl ether

  • MDL number MFCD11113146
  • Cas Number : 69567-11-9
  • Molecular Weight : 523.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-2019

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-2019
Availability details for
Quantity
  • Availability
  • Add to Cart