COMH93D5D72A Combi-Blocks, Inc.

(2-Chloropyridin-3-yl)(4-(hydroxymethyl)piperidin-1-yl)methanone

  • MDL number MFCD11133074
  • Cas Number : 1094301-19-5
  • Molecular Weight : 254.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-2073

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-2073
Availability details for
Quantity
  • Availability
  • Add to Cart