COMH93D5D72B Combi-Blocks, Inc.

4-Amino-1-[4-(hydroxymethyl)piperidin-1-yl]butan-1-one

  • MDL number MFCD11133088
  • Cas Number : 1094389-42-0
  • Molecular Weight : 200.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-2074

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-2074
Availability details for
Quantity
  • Availability
  • Add to Cart