COMH93D5D72E Combi-Blocks, Inc.

N1,N1-Diethyl-4-fluoro-1,2-benzenediamine

  • MDL number MFCD11135621
  • Cas Number : 1019556-91-2
  • Molecular Weight : 182.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-2077

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-2077
Availability details for
Quantity
  • Availability
  • Add to Cart