COMH93D5D74F Combi-Blocks, Inc.

N-(Thiophen-2-ylmethyl)-2-(trifluoromethoxy)aniline

  • MDL number MFCD11143416
  • Cas Number : 1019602-09-5
  • Molecular Weight : 273.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-2111

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-2111
Availability details for
Quantity
  • Availability
  • Add to Cart