COMH93D5D76E Combi-Blocks, Inc.

2-Bromo-1-(cyclopentyloxy)-4-methylbenzene

  • MDL number MFCD11156054
  • Cas Number : 1021142-90-4
  • Molecular Weight : 255.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-2147

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-2147
Availability details for
Quantity
  • Availability
  • Add to Cart