COMH93D5D791 Combi-Blocks, Inc.

Methyl[(4-methylphenyl)methyl](piperidin-4-ylmethyl)amine

  • MDL number MFCD11173249
  • Cas Number : 1830871-65-2
  • Molecular Weight : 232.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-2188

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-2188
Availability details for
Quantity
  • Availability
  • Add to Cart