COMH93D5D7BF Combi-Blocks, Inc.

Methyl 3-amino-3-[3-(trifluoromethyl)phenyl]propanoate

  • MDL number MFCD11200703
  • Cas Number : 1038323-37-3
  • Molecular Weight : 247.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-2239

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-2239
Availability details for
Quantity
  • Availability
  • Add to Cart