COMH93D5D7C5 Combi-Blocks, Inc.

[(3-Fluoro-4-methoxyphenyl)methyl](propan-2-yl)amine

  • MDL number MFCD11204830
  • Cas Number : 1094423-99-0
  • Molecular Weight : 197.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-2246

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-2246
Availability details for
Quantity
  • Availability
  • Add to Cart