COMH93D5D7CE Combi-Blocks, Inc.

7,8-Dimethyl-3,4-dihydro-2h-1-benzopyran-4-one

  • MDL number MFCD11207537
  • Cas Number : 91142-52-8
  • Molecular Weight : 176.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-2256

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-2256
Availability details for
Quantity
  • Availability
  • Add to Cart