COMH93D5D7D7 Combi-Blocks, Inc.

[(6-Methylpyridin-3-yl)methyl](propyl)amine

  • MDL number MFCD11212485
  • Cas Number : 1094511-96-2
  • Molecular Weight : 164.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-2266

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-2266
Availability details for
Quantity
  • Availability
  • Add to Cart