COMH93D5D810 Combi-Blocks, Inc.

1-N-Benzyl-1-n-methyl-2-boc-ethane-1,2-diamine

  • MDL number MFCD11501206
  • Cas Number : 849472-99-7
  • Molecular Weight : 264.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-2333

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-2333
Availability details for
Quantity
  • Availability
  • Add to Cart