COMH93D5D853 Combi-Blocks, Inc.

4-Bromo-2-(2-(dimethylamino)ethoxy)aniline

  • MDL number MFCD11520819
  • Cas Number : 1072906-05-8
  • Molecular Weight : 259.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-2424

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-2424
Availability details for
Quantity
  • Availability
  • Add to Cart