COMH93D5D882 Combi-Blocks, Inc.

2-(Piperidin-4-ylthio)benzo[d]thiazole hydrochloride

  • MDL number MFCD11559911
  • Cas Number : 333992-22-6
  • Molecular Weight : 286.85

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-2482

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-2482
Availability details for
Quantity
  • Availability
  • Add to Cart