COMH93D5D8BF Combi-Blocks, Inc.

Methyl 4-(3-fluorophenyl)-4-oxobutanoate

  • MDL number MFCD11617549
  • Cas Number : 69797-47-3
  • Molecular Weight : 210.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-2554

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-2554
Availability details for
Quantity
  • Availability
  • Add to Cart