COMH93D5D902 Combi-Blocks, Inc.

N-(4-Bromo-2-chlorophenyl)-3-chloropropanamide

  • MDL number MFCD11644505
  • Cas Number : 1098343-13-5
  • Molecular Weight : 296.98

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-2631

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-2631
Availability details for
Quantity
  • Availability
  • Add to Cart