COMH93D5D97C Combi-Blocks, Inc.

2-Bromo-1-(3-(trifluoromethyl)pyridin-2-yl)ethanone

  • MDL number MFCD11847716
  • Cas Number : 749257-79-2
  • Molecular Weight : 268.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-2790

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-2790
Availability details for
Quantity
  • Availability
  • Add to Cart