COMH93D5D97D Combi-Blocks, Inc.

4-Bromopyridine-2-sulfonyl chloride

  • MDL number MFCD11847757
  • Cas Number : 1060808-87-8
  • Molecular Weight : 256.51

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-2791

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-2791
Availability details for
Quantity
  • Availability
  • Add to Cart