COMH93D5DA07 Combi-Blocks, Inc.

1-(3-Chlorophenyl)propan-2-ol

  • MDL number MFCD11934611
  • Cas Number : 343270-59-7
  • Molecular Weight : 170.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-2958

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-2958
Availability details for
Quantity
  • Availability
  • Add to Cart