COMH93D5DA40 Combi-Blocks, Inc.

4,5-Dihydro-2-vinyloxazole

  • MDL number MFCD11976030
  • Cas Number : 13670-33-2
  • Molecular Weight : 97.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-3025

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-3025
Availability details for
Quantity
  • Availability
  • Add to Cart