COMH93D5DA6B Combi-Blocks, Inc.

(S)-Alpha-methyl-2-bromophenylalanine h2o

  • MDL number MFCD11983194
  • Molecular Weight : 276.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-3080

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-3080
Availability details for
Quantity
  • Availability
  • Add to Cart