COMH93D5DA6D Combi-Blocks, Inc.

2-(2-(4-(2-Methoxyphenyl)piperazin-1-yl)ethyl)isoindoline-1,3-dione

  • MDL number MFCD11986896
  • Cas Number : 99718-67-9
  • Molecular Weight : 365.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-3082

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-3082
Availability details for
Quantity
  • Availability
  • Add to Cart