COMH93D5DA73 Combi-Blocks, Inc.

2,2'':7'',2''''-Ter-9,9'-spirobi[9h-fluorene]

  • MDL number MFCD12022449
  • Cas Number : 518997-91-6
  • Molecular Weight : 945.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-3089

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-3089
Availability details for
Quantity
  • Availability
  • Add to Cart