COMH93D5DA81 Combi-Blocks, Inc.

5,6,7,7A-Tetrahydro-5-(triphenylmethyl)thieno[3,2-c]pyridinone

  • MDL number MFCD12024648
  • Cas Number : 109904-26-9
  • Molecular Weight : 397.54

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-3106

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-3106
Availability details for
Quantity
  • Availability
  • Add to Cart