COMH93D5DA8F Combi-Blocks, Inc.

4-(2-Oxoethyl)benzonitrile

  • MDL number MFCD12025076
  • Cas Number : 76113-58-1
  • Molecular Weight : 145.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-3122

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-3122
Availability details for
Quantity
  • Availability
  • Add to Cart