COMH93D5DAD1 Combi-Blocks, Inc.

2-Isopropylpyridin-4-amine

  • MDL number MFCD12032817
  • Cas Number : 340006-70-4
  • Molecular Weight : 136.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-3206

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-3206
Availability details for
Quantity
  • Availability
  • Add to Cart