COMH93D5DAD8 Combi-Blocks, Inc.

[2,2'-Bipyridin]-5-amine

  • MDL number MFCD12033465
  • Cas Number : 160539-04-8
  • Molecular Weight : 171.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-3217

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-3217
Availability details for
Quantity
  • Availability
  • Add to Cart