COMH93D5DB51 Combi-Blocks, Inc.

[(5-Bromo-2-chlorophenyl)methyl](ethyl)amine

  • MDL number MFCD12168573
  • Cas Number : 1096829-54-7
  • Molecular Weight : 248.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-3352

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-3352
Availability details for
Quantity
  • Availability
  • Add to Cart