COMH93D5DB5B Combi-Blocks, Inc.

2-(4-Bromophenyl)-2-hydroxyacetonitrile

  • MDL number MFCD12169330
  • Cas Number : 58289-69-3
  • Molecular Weight : 212.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HB-3364

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HB-3364
Availability details for
Quantity
  • Availability
  • Add to Cart